3',4',5,7-Tetramethoxyflavone, 97%
3',4',5,7-Tetramethoxyflavone, 97%
3',4',5,7-Tetramethoxyflavone, 97%
Thermo Scientific Chemicals

3',4',5,7-Tetramethoxyflavone, 97%

CAS: 855-97-0 | C19H18O6 | 342.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number L14496.MC
also known as L14496-MC
Price (EUR)
64,65
キャンペーン価格
71,90
Save 7,25 (10%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (EUR)
64,65
キャンペーン価格
71,90
Save 7,25 (10%)
Each
Chemical Identifiers
CAS855-97-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Melting Point (clear melt)191.5-200.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only