1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine, 97%
1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine, 97%
1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine, 97%
Thermo Scientific Chemicals

1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine, 97%

CAS: 202823-67-4 | C10H10Cl2N2 | 229.104 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L14835.03
also known as L14835-03
Price (EUR)
58,50
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
58,50
Each
Chemical Identifiers
CAS202823-67-4
IUPAC Name3,6-dichloro-4,5-diazatricyclo[6.2.2.0²,⁷]dodeca-2(7),3,5-triene
Molecular FormulaC10H10Cl2N2
InChI KeyGZIFYLGNTOAIJJ-UHFFFAOYSA-N
SMILESClC1=NN=C(Cl)C2=C1C1CCC2CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Melting Point (clear melt)207.5-216.5?C
1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only