Thermo Scientific Chemicals

1-Methyl-1-cyclohexene, 96%, Thermo Scientific Chemicals

Catalog number: L15014.06
5 g, Each
Thermo Scientific Chemicals

1-Methyl-1-cyclohexene, 96%, Thermo Scientific Chemicals

Catalog number: L15014.06
5 g, Each
Quantity
Catalog number: L15014.06
also known as L15014-06
Price (EUR)
Price: 25,70
Online price: 23,13
Your price:
Quantity
-

Chemical Identifiers

CAS
591-49-1
IUPAC Name
1-methylcyclohex-1-ene
Molecular Formula
C7H12
InChI Key
CTMHWPIWNRWQEG-UHFFFAOYSA-N
SMILES
CC1=CCCCC1
Appearance (Color)
Clear, colorless
Assay (GC)
>95.0%
Comment
Material sourced in the U.S. and in other countries
Refractive Index
1.4420-1.4520 @ 20°C (non-U.S. sourced material)
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 2
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Aspiration Toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H304 - May be fatal if swallowed and enters airways

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Physical Hazards

    H225 - Highly flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P308 + P313 - IF exposed or concerned: Get medical advice/attention