3-Iodophenylacetic acid, 98%
3-Iodophenylacetic acid, 98%
3-Iodophenylacetic acid, 98%
Thermo Scientific Chemicals

3-Iodophenylacetic acid, 98%

CAS: 1878-69-9 | C8H7IO2 | 262.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2 g
Catalog number L15117.ME
also known as L15117-ME
Price (EUR)
40,10
Each
Quantity:
500 mg
Request bulk or custom format
Price (EUR)
40,10
Each
Chemical Identifiers
CAS1878-69-9
IUPAC Name2-(3-iodophenyl)acetic acid
Molecular FormulaC8H7IO2
InChI KeyMRSWWBAFGGGWRH-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC(I)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Melting Point (clear melt)128.0-134.0?C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only