Pyrrole-2-carbonitrile, 99%
Pyrrole-2-carbonitrile, 99%
Pyrrole-2-carbonitrile, 99%
Thermo Scientific Chemicals

Pyrrole-2-carbonitrile, 99%

CAS: 4513-94-4 | C5H4N2 | 92.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L15330.03
also known as L15330-03
Price (EUR)
47,80
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
47,80
Each
Chemical Identifiers
CAS4513-94-4
IUPAC Name1H-pyrrole-2-carbonitrile
Molecular FormulaC5H4N2
InChI KeyBQMPGKPTOHKYHS-UHFFFAOYSA-N
SMILESN#CC1=CC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>98.5%
Refractive Index1.5475-1.5525 @ 20?C
Appearance (Color)Clear, colorless to yellow
FormLiquid
Pyrrole-2-carbonitrile is useful in addition reactions and in heterocycle formation. It acts as a heterocyclic building block in synthetic chemistry. Further, it is used in the preparation of 4-iodopyrrole-2-carbonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrole-2-carbonitrile is useful in addition reactions and in heterocycle formation. It acts as a heterocyclic building block in synthetic chemistry. Further, it is used in the preparation of 4-iodopyrrole-2-carbonitrile.

Solubility
Miscible with benzene, dichloromethane, tertrahydrofuran, dimethylformamide and acetonitrile.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

General References:

  1. Seo, S.; DeSilva, M. A.; Xia, H.; Brennecke, J. F. Effect of Cation on Physical Properties and CO2 Solubility for Phosphonium-Based Ionic Liquids with 2-Cyanopyrrolide Anions. J. Phys. Chem. B 2015, 119 (35), 11807-11814.
  2. Sun, L.; Morales-Collazo, O.; Xia, H.; Brennecke, J. F. Effect of Structure on Transport Properties (Viscosity, Ionic Conductivity, and Self-Diffusion Coefficient) of Aprotic Heterocyclic Anion (AHA) Room-Temperature Ionic Liquids. 1. Variation of Anionic Species. J. Phys. Chem. B 2015, 119 (48), 15030-15039.