3-Indolemethanol, 97%
3-Indolemethanol, 97%
3-Indolemethanol, 97%
Thermo Scientific Chemicals

3-Indolemethanol, 97%

CAS: 700-06-1 | C9H9NO | 147.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L15762.03
also known as L15762-03
Price (EUR)
25,10
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
25,10
Each
Chemical Identifiers
CAS700-06-1
IUPAC Name(1H-indol-3-yl)methanol
Molecular FormulaC9H9NO
InChI KeyIVYPNXXAYMYVSP-UHFFFAOYSA-N
SMILESOCC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to dark cream to cream to orange to pale brown
FormCrystals or powder or crystalline powder or flakes
Assay (Silylated GC)≥96.0%
Melting Point (clear melt)93.0-102.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only