Chemical Identifiers
CAS141607-35-4
IUPAC Name6-{[(tert-butyldiphenylsilyl)oxy]methyl}-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-ol
Molecular FormulaC24H32O5Si
InChI KeyOOVHEXLXHQNNLL-UHFFFAOYNA-N
SMILESCC(C)(C)[Si](OCC1OC(O)C2OC(C)(C)OC12)(C1=CC=CC=C1)C1=CC=CC=C1
View more