2,5-Difluorophenyl isocyanate, 98%
2,5-Difluorophenyl isocyanate, 98%
2,5-Difluorophenyl isocyanate, 98%
2,5-Difluorophenyl isocyanate, 98%
Thermo Scientific Chemicals

2,5-Difluorophenyl isocyanate, 98%

CAS: 39718-32-6 | C7H3F2NO | 155.104 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L15996.03
also known as L15996-03
Price (EUR)
40,80
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
40,80
Each
Add to cart
Chemical Identifiers
CAS39718-32-6
IUPAC Name1,4-difluoro-2-isocyanatobenzene
Molecular FormulaC7H3F2NO
InChI KeySNHIIFOXCRYGGY-UHFFFAOYSA-N
SMILESFC1=CC=C(F)C(=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4925-1.4965 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only