Amberlite XAD-1180
Thermo Scientific Chemicals

Amberlite XAD-1180

CAS: 97396-56-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number L16097.22
also known as L16097-22
Price (EUR)
37,10
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
37,10
Each
Chemical Identifiers
CAS97396-56-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormBeads
CommentUniformity coefficient: 0.00-1.80
Harmonic mean size0.350-0.600 mm
Particle Size≤0.250mm: 0-5%
View more
Polymeric adsorbent for hydrophobic compounds: antibiotics, vitamins, amino acids, enzymes; for purification: gallium, myxovirescins, phosphoric acid, riboflavin salts. Amberlite™ XAD1180N can be used as a polymeric adsorbent. The regeneration of this adsorbent has been effective in alkaline medium and tests have shown the adsorbent efficiently removes chromium from tannery waste water. Mainly used in the Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polymeric adsorbent for hydrophobic compounds: antibiotics, vitamins, amino acids, enzymes; for purification: gallium, myxovirescins, phosphoric acid, riboflavin salts. Amberlite™ XAD1180N can be used as a polymeric adsorbent. The regeneration of this adsorbent has been effective in alkaline medium and tests have shown the adsorbent efficiently removes chromium from tannery waste water. Mainly used in the Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. A.S., et al. An enhanced adsorption methodology for the detoxification of chromium using n-octylamine impregnated Amberlite XAD-4 polymeric sorbent Kumar. J. Environ. Sci. Health. 2011, 461598-1610.
  2. Xiaowei Zeng et. al. Enhanced adsorption of phenol from water by a novel polar post-crosslinked polymeric adsorbent. Journal of Hazardous Materials.2009, 169 (1-3), 1022-1028 .
  3. Non-ionic polystyrene adsorbent, minimum surface area 550 m2/g, 20-60 mesh.