4-Amino-2-isopropyl-5-methylphenol hydrochloride, 97%
4-Amino-2-isopropyl-5-methylphenol hydrochloride, 97%
4-Amino-2-isopropyl-5-methylphenol hydrochloride, 97%
Thermo Scientific Chemicals

4-Amino-2-isopropyl-5-methylphenol hydrochloride, 97%

CAS: 6321-11-5 | C6H10O6 | 178.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L16282.03
also known as L16282-03
Price (EUR)
22,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
22,30
Each
Chemical Identifiers
CAS6321-11-5
IUPAC Name4-amino-5-methyl-2-(propan-2-yl)phenol hydrochloridyl
Molecular FormulaC10H15ClNO
InChI KeyYLWORLAQZBRLSS-UHFFFAOYSA-N
SMILES[Cl].CC(C)C1=CC(N)=C(C)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Appearance (Color)Cream to pink or yellow to brown
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only