3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane, 98%
3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane, 98%
3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane, 98%
Thermo Scientific Chemicals

3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane, 98%

CAS: 17096-07-0 | C16H38O5Si4 | 422.815 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L16682.22
also known as L16682-22
Price (EUR)
298,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
298,00
Each
Chemical Identifiers
CAS17096-07-0
IUPAC Name3-{2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptan-4-yl}propyl 2-methylprop-2-enoate
Molecular FormulaC16H38O5Si4
InChI KeyBESKSSIEODQWBP-UHFFFAOYSA-N
SMILESCC(=C)C(=O)OCCC[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4175-1.4215 @ 20?C
3-(Methacryloyloxy)propyltris(trimethylsiloxy) silane acts as a tetramethylsilane-protected silicate compound utilized for proteomics research. It is also used as a silane coupling agent, silane adhesion promoter, silane hydrophobing agent, silane dispersing agent and silane crosslinking agent. It is also employed as a silane moisture scavenger, polypropylene catalyst, silicate stabilizer, polyurethane endcapper silane, silane drying agent, silane curing agent and silane reinforcer. Further, it serves as a silylating agent, silane reducing agent, silyl building block and synthons in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Methacryloyloxy)propyltris(trimethylsiloxy) silane acts as a tetramethylsilane-protected silicate compound utilized for proteomics research. It is also used as a silane coupling agent, silane adhesion promoter, silane hydrophobing agent, silane dispersing agent and silane crosslinking agent. It is also employed as a silane moisture scavenger, polypropylene catalyst, silicate stabilizer, polyurethane endcapper silane, silane drying agent, silane curing agent and silane reinforcer. Further, it serves as a silylating agent, silane reducing agent, silyl building block and synthons in synthetic chemistry.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agent.
RUO – Research Use Only

General References:

  1. Demianenko, P.; Minisini, B.; Lamrani, M.; Poncin-Epaillard, F. Synthesis and characterization of simultaneous IPNs. Evidence of the IPN structure by selective chemical attack on the cross-linker. Mater. Today Commun. 2015, 4, 1-5.
  2. Guisasola, E.; Baeza, A.; Talelli, M.; Arcos, D.; Moros, M.; de la Fuente, J. M.; Vallet-Regí, M. Magnetic-Responsive Release Controlled by Hot Spot Effect. Langmuir 2015, 31 (46), 12777-12782.