Ultramark∣r 1621, Mass Spec Std
Ultramark∣r 1621, Mass Spec Std
Ultramark∣r 1621, Mass Spec Std
Ultramark∣r 1621, Mass Spec Std
Thermo Scientific Chemicals

Ultramark∣r 1621, Mass Spec Std

CAS: 105809-15-2 | C6H18N3P3 | 231.189 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L16698.MD
also known as L16698-MD
Price (EUR)
47,20
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
47,20
Each
Add to cart
Chemical Identifiers
CAS105809-15-2
IUPAC Namehexamethyl-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine
Molecular FormulaC6H18N3P3
InChI KeyDEBZEVJNWCNATM-UHFFFAOYSA-N
SMILESCP1(C)=NP(C)(C)=NP(C)(C)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear or slightly hazy, yellow
FormLiquid
Assay from Suppliers CofA≥95%
GC/MS AnalysisConforms to reference
Ultramark 1621 is used as a reference compound for positive and negative ion fast-atom bombardment high-resolution mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ultramark 1621 is used as a reference compound for positive and negative ion fast-atom bombardment high-resolution mass spectrometry.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Fluorinated phosphazine for mass spectrometry showing peaks up to 2120 mass units.
  2. Seró, R.; Núñez, O.; Bosch, J.; Grases, J. M.; Rodríguez, P.; Moyano, E.; Galceran, M. T. Desorption electrospray ionization-high resolution mass spectrometry for the screening of veterinary drugs in cross-contaminated feedstuffs. Anal. Bioanal. Chem. 2015, 407 (24), 7369-7378.
  3. Gómez-Pérez, M. L.; Romero-González, R.; Martínez, V. J. L.; Frenich, A. G. Analysis of pesticide and veterinary drug residues in baby food by liquid chromatography coupled to Orbitrap high resolution mass spectrometry. Talanta 2015, 131, 1-7.