2,2,2-Trifluoroethanol, NMR grade, 99.5%
2,2,2-Trifluoroethanol, NMR grade, 99.5%
2,2,2-Trifluoroethanol, NMR grade, 99.5%
Thermo Scientific Chemicals

2,2,2-Trifluoroethanol, NMR grade, 99.5%

CAS: 75-89-8 | C2H3F3O | 100.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L16898.22
also known as L16898-22
Price (EUR)
73,65
Online exclusive
81,60
Save 7,95 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
73,65
Online exclusive
81,60
Save 7,95 (10%)
Each
Chemical Identifiers
CAS75-89-8
IUPAC Name2,2,2-trifluoroethan-1-ol
Molecular FormulaC2H3F3O
InChI KeyRHQDFWAXVIIEBN-UHFFFAOYSA-N
SMILESOCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear, colorless
Assay (GC)≥99.5%
Refractive Index<1.3000 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only