2-(Hydroxymethyl)tetrahydropyran, 94%
2-(Hydroxymethyl)tetrahydropyran, 94%
2-(Hydroxymethyl)tetrahydropyran, 94%
Thermo Scientific Chemicals

2-(Hydroxymethyl)tetrahydropyran, 94%

CAS: 100-72-1 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L17417.06
also known as L17417-06
Price (EUR)
36,70
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
36,70
Each
Chemical Identifiers
CAS100-72-1
IUPAC Name(oxan-2-yl)methanol
Molecular FormulaC6H12O2
InChI KeyROTONRWJLXYJBD-UHFFFAOYNA-N
SMILESOCC1CCCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥92.0%
Refractive Index1.4535-1.4605 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only