3-Cyano-5-methylpyridine, 98+%
3-Cyano-5-methylpyridine, 98+%
3-Cyano-5-methylpyridine, 98+%
Thermo Scientific Chemicals

3-Cyano-5-methylpyridine, 98+%

CAS: 42885-14-3 | C7H6N2 | 118.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L17427.03
also known as L17427-03
Price (EUR)
79,40
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
79,40
Each
Specifications
Beilstein111258
CAS42885-14-3
Chemical Name or Material3-Cyano-5-methylpyridine
EINECS Number255-988-8
Formula Weight118.14
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only