1-Isopropenyl-2-benzimidazolidinone, 98+%
1-Isopropenyl-2-benzimidazolidinone, 98+%
1-Isopropenyl-2-benzimidazolidinone, 98+%
Thermo Scientific Chemicals

1-Isopropenyl-2-benzimidazolidinone, 98+%

CAS: 52099-72-6 | C10H10N2O | 174.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17626.03
also known as L17626-03
Price (EUR)
42,10
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
42,10
Each
Chemical Identifiers
CAS52099-72-6
IUPAC Name1-(prop-1-en-2-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one
Molecular FormulaC10H10N2O
InChI KeyXFASJWLBXHWUMW-UHFFFAOYSA-N
SMILESCC(=C)N1C(=O)NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
Melting Point (clear melt)119.0-125.0?C
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only