3-Iodobenzylamine, 97%
3-Iodobenzylamine, 97%
3-Iodobenzylamine, 97%
Thermo Scientific Chemicals

3-Iodobenzylamine, 97%

CAS: 696-40-2 | C7H8IN | 233.052 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17801.03
also known as L17801-03
Price (EUR)
112,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
112,00
Each
Chemical Identifiers
CAS696-40-2
IUPAC Name1-(3-iodophenyl)methanamine
Molecular FormulaC7H8IN
InChI KeyLQLOGZQVKUNBRX-UHFFFAOYSA-N
SMILESNCC1=CC=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to white to yellow
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥96.0%
Refractive Index1.6355-1.6405 @ 20?C
It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Solubility
Difficult to mix in water.

Notes
Air sensitive and light sensitive. Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kenichi Akaji.; Yoshiaki Kiso. Macrocyclization on solid support using Heck reaction.Tetrahedron Lett. 1997, 38 (29),5185-5188 .
  2. Jeffrey B Arterburn.; Kalla Venkateswara Rao.; Marc C Perry. Novel 17α-ethynylestradiol derivatives: Sonogashira couplings using unprotected phenylhydrazines.Tetrahedron Lett. 2000, 41 (6),839-842 .