3-Formyl-4-methoxybenzeneboronic acid, 98%
3-Formyl-4-methoxybenzeneboronic acid, 98%
3-Formyl-4-methoxybenzeneboronic acid, 98%
Thermo Scientific Chemicals

3-Formyl-4-methoxybenzeneboronic acid, 98%

CAS: 121124-97-8 | C8H9BO4 | 179.966 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17850.03
also known as L17850-03
Price (EUR)
26,50
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
26,50
Each
Chemical Identifiers
CAS121124-97-8
IUPAC Name(3-formyl-4-methoxyphenyl)boronic acid
Molecular FormulaC8H9BO4
InChI KeyYJQDBKGGRPJSOI-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1C=O)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMRConforms to structure
Assay (Aqueous acid-base Titration)≥97.5%
Appearance (Color)White to cream
Assay (HPLC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only