(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine, 97%, Thermo Scientific Chemicals
(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine, 97%, Thermo Scientific Chemicals
(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L18659.03
also known as L18659-03
1 g
Catalog number L18659.03
also known as L18659-03
Price (EUR)
170,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
170,00
Each
Chemical Identifiers
CAS153837-28-6
IUPAC Name(2,2-dimethylpropyl)[1-phenyl-2-(piperidin-1-yl)ethyl]amine
Molecular FormulaC18H30N2
InChI KeyRUWFXOINQANLGF-UHFFFAOYNA-N
SMILESCC(C)(C)CNC(CN1CCCCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused solid or cloudy melt
Optical Rotation-92? ±3? (c=2 in methanol)
Appearance (Color)White to yellow
Melting Point36-46?C
Assay (GC)≥96.0%
(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine is a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Store under inert gas. Incompatible with oxidizing agents.
RUO – Research Use Only