3-(Methoxycarbonyl)phenyl isocyanate, 97%
3-(Methoxycarbonyl)phenyl isocyanate, 97%
3-(Methoxycarbonyl)phenyl isocyanate, 97%
Thermo Scientific Chemicals

3-(Methoxycarbonyl)phenyl isocyanate, 97%

CAS: 41221-47-0 | C9H7NO3 | 177.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L18709.03
also known as L18709-03
Price (EUR)
55,65
キャンペーン価格
62,10
Save 6,45 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
55,65
キャンペーン価格
62,10
Save 6,45 (10%)
Each
Chemical Identifiers
CAS41221-47-0
IUPAC Namemethyl 3-isocyanatobenzoate
Molecular FormulaC9H7NO3
InChI KeyWBGWGERFPSYHDT-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
Assay (GC)≥96.0%
Melting Point31.0-41.0°C
CommentMelting point may be hazy
FormCrystals or powder or crystalline powder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only