4-Nitrophenylketoxime on polystrene, 2% cross-linked, 200-400 mesh, 0.8-1.0 mmol/g
4-Nitrophenylketoxime on polystrene, 2% cross-linked, 200-400 mesh, 0.8-1.0 mmol/g
4-Nitrophenylketoxime on polystrene, 2% cross-linked, 200-400 mesh, 0.8-1.0 mmol/g
Thermo Scientific Chemicals

4-Nitrophenylketoxime on polystrene, 2% cross-linked, 200-400 mesh, 0.8-1.0 mmol/g

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19376.03
also known as L19376-03
Price (EUR)
44,20
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
44,20
Each
Specifications
Chemical Name or Material4-Nitrophenylketoxime on polystrene
Concentration or Composition (by Analyte or Components)0.8 to 1.0mmol/g
MDL NumberMFCD00165078
Mesh Size200-400 Mesh
Name Note2% cross-linked
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only