3-Fluoro-4-methylbenzenesulfonyl chloride, 97%
3-Fluoro-4-methylbenzenesulfonyl chloride, 97%
3-Fluoro-4-methylbenzenesulfonyl chloride, 97%
3-Fluoro-4-methylbenzenesulfonyl chloride, 97%
Thermo Scientific Chemicals

3-Fluoro-4-methylbenzenesulfonyl chloride, 97%

CAS: 90260-13-2 | C7H6ClFO2S | 208.631 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19516.06
also known as L19516-06
Price (EUR)
132,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
132,00
Each
Add to cart
Chemical Identifiers
CAS90260-13-2
IUPAC Name3-fluoro-4-methylbenzene-1-sulfonyl chloride
Molecular FormulaC7H6ClFO2S
InChI KeyYDUHIMCRFRIVFI-UHFFFAOYSA-N
SMILESCC1=C(F)C=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)44.5-53.5?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only