(R)-(-)-1-Methyl-3-hydroxypyrrolidine, 99%
(R)-(-)-1-Methyl-3-hydroxypyrrolidine, 99%
(R)-(-)-1-Methyl-3-hydroxypyrrolidine, 99%
Thermo Scientific Chemicals

(R)-(-)-1-Methyl-3-hydroxypyrrolidine, 99%

CAS: 104641-60-3 | C5H11NO | 101.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19694.03
also known as L19694-03
Price (EUR)
76,50
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
76,50
Each
Chemical Identifiers
CAS104641-60-3
IUPAC Name1-methylpyrrolidin-3-ol
Molecular FormulaC5H11NO
InChI KeyFLVFPAIGVBQGET-UHFFFAOYNA-N
SMILESCN1CCC(O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange to brown
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Optical Rotation-7.0? ± 1.0? (c=1 in chloroform)
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only