2-Amino-3,5-difluoropyridine, 98%, Thermo Scientific Chemicals
2-Amino-3,5-difluoropyridine, 98%, Thermo Scientific Chemicals
2-Amino-3,5-difluoropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-3,5-difluoropyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L19727.06
also known as L19727-06
5 g
Catalog number L19727.06
also known as L19727-06
Price (EUR)
86,10
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
86,10
Each
Chemical Identifiers
CAS732306-31-9
IUPAC Name3,5-difluoropyridin-2-amine
Molecular FormulaC5H4F2N2
InChI KeyJVLFMTZUPSBCNJ-UHFFFAOYSA-N
SMILESNC1=NC=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Appearance (Color)White to pale brown
Melting Point (clear melt)54.0-60.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only