2,2-Dibromo-4'-fluoroacetophenone, 98%
2,2-Dibromo-4'-fluoroacetophenone, 98%
2,2-Dibromo-4'-fluoroacetophenone, 98%
Thermo Scientific Chemicals

2,2-Dibromo-4'-fluoroacetophenone, 98%

CAS: 7542-64-5 | C8H5Br2FO | 295.933 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19788.03
also known as L19788-03
Price (EUR)
12,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
12,30
Each
Chemical Identifiers
CAS7542-64-5
IUPAC Name2,2-dibromo-1-(4-fluorophenyl)ethan-1-one
Molecular FormulaC8H5Br2FO
InChI KeyXFFGHBAVJPOWDG-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)C(=O)C(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)34.0-40.0?C
2,2-Dibromo-4'-fluoroacetophenone is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dibromo-4′-fluoroacetophenone is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
RUO – Research Use Only