N-Boc-1,6-diaminohexane, 98%
N-Boc-1,6-diaminohexane, 98%
N-Boc-1,6-diaminohexane, 98%
Thermo Scientific Chemicals

N-Boc-1,6-diaminohexane, 98%

CAS: 51857-17-1 | C11H24N2O2 | 216.325 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L19948.03
also known as L19948-03
Price (EUR)
48,80
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
48,80
Each
Chemical Identifiers
CAS51857-17-1
IUPAC Nametert-butyl N-(6-azaniumylhexyl)carbamate
Molecular FormulaC11H25N2O2
InChI KeyRVZPDKXEHIRFPM-UHFFFAOYSA-O
SMILESCC(C)(C)OC(=O)NCCCCCC[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4585-1.4625 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid or viscous liquid
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
N-Boc-1,6-diaminohexane is used to prepare 1,3-Bis[6-(Boc-amino)hexyl]urea by reacting with carbonyl dichloride in the presence of triethylamine. Further, it is used as a reagent for the introduction of a C6-spacer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-1,6-diaminohexane is used to prepare 1,3-Bis[6-(Boc-amino)hexyl]urea by reacting with carbonyl dichloride in the presence of triethylamine. Further, it is used as a reagent for the introduction of a C6-spacer.

Solubility
Soluble in dichloromethane and ethyl acetate.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Jong, T.; Bradley, M. Flow-Mediated Synthesis of Boc, Fmoc, and Ddiv Monoprotected Diamines. Org. Lett. 2015, 17 (3), 422-425.
  2. Jong, T.; Pérez-López, A. M.; Johansson, E. M. V.; Lilienkampf, A.; Bradley, M. Flow and Microwave-Assisted Synthesis of N-(Triethylene glycol)glycine Oligomers and Their Remarkable Cellular Transporter Activities. Bioconjugate Chem. 2015, 26 (8), 1759-1765.