2-Nitrophenylselenocyanate, 97%
2-Nitrophenylselenocyanate, 97%
2-Nitrophenylselenocyanate, 97%
Thermo Scientific Chemicals

2-Nitrophenylselenocyanate, 97%

CAS: 51694-22-5 | C7H4N2O2Se | 227.092 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20193.03
also known as L20193-03
Price (EUR)
53,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
53,00
Each
Chemical Identifiers
CAS51694-22-5
IUPAC Name[(2-nitrophenyl)selanyl]formonitrile
Molecular FormulaC7H4N2O2Se
InChI KeyLHBLJWULWKQRON-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1[Se]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)134-146?C
Appearance (Color)Brown
Assay (GC)≥96.0%
Identification (FTIR)Conforms
FormPowder or crystalline powder
2-Nitrophenylselenocyanate is used to study the mechanism of its reaction with the zinc/thiolate clusters of metallothionein. It is also used in the preparation of 2,3-seco-5 alfa-cholestane-2,3-diol and in the preparation of 4alfa-methyl-2,3-seco-5 alfa-cholestane-2,3-diol. Further, it is used in the preparation of 2-nitrophenylselenyl derivative and 3alfa-[(2-Nitrophenyl)seleno]androsta-1,5-dien-17beta-ol. In addition to this, it reacts with 6-bromo-hexanoic acid methyl ester to get 6-(2-nitro-phenylselanyl)-hexanoic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrophenylselenocyanate is used to study the mechanism of its reaction with the zinc/thiolate clusters of metallothionein. It is also used in the preparation of 2,3-seco-5 alfa-cholestane-2,3-diol and in the preparation of 4alfa-methyl-2,3-seco-5 alfa-cholestane-2,3-diol. Further, it is used in the preparation of 2-nitrophenylselenyl derivative and 3alfa-[(2-Nitrophenyl)seleno]androsta-1,5-dien-17beta-ol. In addition to this, it reacts with 6-bromo-hexanoic acid methyl ester to get 6-(2-nitro-phenylselanyl)-hexanoic acid methyl ester.

Solubility
Soluble in dichloromethane, tetrahydrofuran, dimethyl formamide and ethanol.

Notes
Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kashinath, K.; Dhara, S.; Reddy, D. S. Breaking and Making of Olefins Simultaneously Using Ozonolysis: Application to the Synthesis of Useful Building Blocks and Macrocyclic Core of Solomonamides. Org. Lett. 2015, 17 (9), 2090-2093.
  2. Bender, M.; Schmidtmann, M.; Summons, R. E.; Rullkötter, J.; Christoffers, J. A Geomimetic Approach to the Formation and Identification of Fossil Sterane Biomarkers in Crude Oil: 18-nor-D-homo-Androstane and 5 alpha,14 beta-Androstane. Chem. Eur. J. 2015, 21 (35), 12501-12508.