Blood Agar Base No.2 with Horse Blood
Thermo Scientific™

Blood Agar Base No.2 with Horse Blood

Cultivate and isolate fastidious pathogens and other microorganisms with clearly visible hemolytic reactions with this ready-to-use medium.
Have Questions?
Catalog NumberDescription
PB0114ABlood Agar No. 2 with Horse Blood
Catalog number PB0114A
Price (EUR)
-
Description:
Blood Agar No. 2 with Horse Blood

Cultivate and isolate fastidious pathogens and other microorganisms with clearly visible hemolytic reactions with Thermo Scientific™ Oxoid™ Blood Agar Base No.2 with Horse Blood. The specially designed formulation meets the need for a nutritious blood agar which permits maximum recovery of delicate organisms without interfering with their hemolytic reactions. Added defibrinated horse blood provides additional growth factors and enables the determination of hemolytic patterns.

Use Blood Agar Base No. 2 with Horse Blood for the recovery of fastidious organisms without interfering with their hemolytic reactions. The medium utilizes Thermo Scientific™Oxoid™ Blood Agar Base No.2 as a base medium.

  • Exhibits superior growth promoting properties and enhanced pigment formation in chromogenic bacteria.
  • Supports luxuriant early colonial development of fastidious organisms, particularly streptococci and pneumococci.
  • Allows determination of hemolytic reaction patterns with the addition of defibrinated horse blood.
  • Ready-to-use convenience of prepared media.

In comparison with fresh digest agar, Blood Agar Base No.2 shows equal or superior growth promoting properties and enhanced pigment formation in bacteria. Comparison with many other blood agars has shown that with Oxoid Blood Agar Base No.2 growth of many bacteria is considerably improved, especially the fastidious streptococci and pneumococci, as demontrated by luxuriant and early colonial development.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
ColorRed
DescriptionBlood Agar No. 2 with Horse Blood
Fill Weight19.5g±1.0.g
Format90mm Monoplate
Packaging Type10 x 90mm plates, film wrapped
Quantity10/Pk.
Shelf Life6 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2°C to 10°C