Staphylococci Streptococci Selective Medium/Sabouraud Glucose Selective Agar with Chloramphenicol Biplate
Thermo Scientific™
Staphylococci Streptococci Selective Medium/Sabouraud Glucose Selective Agar with Chloramphenicol Biplate
Isolate staphylococci and streptococci while simultaneously isolating dermatophytes, other fungi and yeasts using this convenient bi-plate.
Have Questions?
Catalog number PB1219E
Price (EUR)
-
Isolate staphylococci and streptococci while simultaneously isolating dermatophytes, other fungi and yeasts from clinical, environmental and food samples using Thermo Scientific™ Oxoid™ Staphylococci Streptococci Selective Medium/Sabouraud Glucose Selective Agar with Chloramphenicol Biplate.
Staphylococci Streptococci Selective Medium ensures clear isolation of staphylococci and streptococci, while Oxoid Sabouraud Glucose Selective Agar provides isolation of pathogenic fungi. The convenient biplate format saves space in automated specimen processing to achieve greater throughput.
Produce more diagnostic details of pathogenic organisms while reducing storage capacity issue in the incubator with a bi-plate that allows simultaneous isolation of staphylococci, streptococci and pathogenic fungi
  • Suitable for identification of pathogenic fungi from a material containing large numbers of other fungi or bacteria
  • Ready-to-use convenience of prepared media.
  • Space-saving bi-plate format.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
ColorCarmine red, opaque/, Ivory, transparent
DescriptionStaphylococci Streptococci Selective Medium/ Sabouraudglucose Selective Agar with Chloramphenicol Biplate
Fill Weight17g ± 5 %
FormatBiplate
Packaging TypeFlow Wrapped - Single Layer
Quantity10 Pack
Shelf Life8 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2°C to 8°C