Columbia Chocolate Agar/MacConkey Agar with Salt Biplate
Thermo Scientific™
Columbia Chocolate Agar/MacConkey Agar with Salt Biplate
Cultivate, isolate and identify a broad range of microorganisms while isolating and differentiating coliforms and intestinal pathogens with this convenient bi-plate.
Have Questions?
Catalog number PB1262E
Price (EUR)
-
Cultivate, isolate and identify a broad range fastidious organisms while isolating and differentiating coliforms and intestinal pathogens in water, dairy and biological samples with Thermo Scientific™ Oxoid™ Columbia Chocolate Agar/MacConkey Agar with Salt Biplate.
Columbia Chocolate Agar facilitates isolation and cultivation of fastidious organisms while MacConkey Agar with Salt is a differential medium for the detection and isolation of pathogens. MacConkey Agar with Salt also provides a number of diagnostic differentiators such as bile tolerance, colony morphology, and chromogenesis.
Read more results from a single patient sample and produce more diagnostic details of pathogenic organisms while reducing storage capacity issues in the incubator with a bi-plate that allows the detection and isolation of Gram positive and Gram negative pathogens on a single plate facilitating faster diagnostic decision making.
  • Cultivate pathogens from clinical specimens and dairy products.
  • Ready-to-use convenience of prepared media
  • Space-saving bi-plate format
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
ColorFawn brown, transparent /Salmon pink, transparen
DescriptionColumbia Chocolate Agar/MacConkey Agar with Salt Biplate
Fill Weight17g ± 5 %
FormatBiplate
Packaging TypeFlow Wrapped - Single Layer
Quantity10 Pack
Shelf Life8 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar Plate
Unit SizeEach
Contents & Storage
2°C to 8°C