R2 A Agar
R2 A Agar
Thermo Scientific™

R2 A Agar

Enumerate heterotrophic bacteria from drinking water samples with this ready-to-use medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
PO0659AStraw, clear
PO5149AOyster white, transparent
Catalog number PO0659A
Price (EUR)
-
Request A Quote
Color:
Straw, clear

Enumerate heterotrophic bacteria from drinking water samples with ready-to-use Thermo Scientific™ Oxoid™ R2A Agar. This nutritionally reduced medium was specially designed to enhance the recovery of stressed and chlorine damaged bacteria from treated water samples.

Use R2A Agar for the microbiological examination of drinking water.

  • Aids recovery of stressed and damaged bacteria giving more realistic colony count as compared to traditional media.
  • Ready-to-use convenience of prepared media.

Standard methods for the enumeration of heterotrophic bacteria in water have traditionally used nutritionally rich media, such as Plate Count Agar, with incubation at 35°C1. Organisms isolated under these conditions may represent only a small percentage of the bacteria present in the sample2.

R2A Agar developed by Reasoner and Geldreich2 is a nutritionally reduced medium. It was demonstrated that using this medium and incubating for longer at lower temperatures enhanced the recovery of stressed and chlorine damaged bacteria from treated waters resulting in higher, more realistic bacterial counts.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

General References:

  1. Greenbreg, Trussell and Clesceri (ed) (1998) Standard Methods for the Examination of Drinking Water and Waste Water. 20th Ed. APHA, Washington DC.
  2. Reasoner and Geldreich (1985). Appl. Environ. Microbiol. 49, 1.
Specifications
ColorStraw, clear
DescriptionR2A Agar
Fill Weight18.5g – 20.5g
Format90mm Plate
Packaging TypeFlow Wrapped - Single Layer
Quantity10/Pk.
Shelf Life9 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach