Chromogenic Listeria Agar (ISO)
Actual product may vary
Thermo Scientific™

Chromogenic Listeria Agar (ISO)

Detect Listeria monocytogenes from food samples with this special formulation which produces an opaque white halo around the colony and conforms to ISO 11290-1 and 11290-2.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatMedia TypeManufacturing Origin
PO1196A10/Pk.90mm MonoplateCLA (ISO)United Kingdom
PO5183A10/Pk.90mm MonoplateCLA (ISO)Germany
Catalog number PO1196A
Price (EUR)
-
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
CLA (ISO)
Manufacturing Origin:
United Kingdom
Isolate and detect Listeria monocytogenes from food samples using Thermo Scientific™ Chromogenic Listeria Agar (ISO), a specially formulated media which conforms to ISO 11290-1 and 11290-2. The expression of the phospholipase C enzyme produced by L. monocytogenes and Listeria ivanovii creates a unique opaque white halo around the L. mono colony which is the diagnostic criteria necessary to differentiate from non-pathogenic Listeria spp.
Use this medium for the isolation, enumeration and presumptive identification of Listeria spp.

This product may not be available in all countries or territories. Please consult your local representative for availability.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
For Laboratory Use
For Laboratory Use
Specifications
Certifications/ComplianceAccording ISO 11290-1 and ISO 11290-2
ColorStraw, semi-opaque
Fill Weight19.5g ± 1.0g
Format90mm Monoplate
Media TypeCLA (ISO)
Packaging TypeFlow Wrapped - Single Layer
Quantity10/Pk.
Shelf Life10 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2°C to 10°C