Gassner Medium
Thermo Scientific™

Gassner Medium

Detect and isolate Enterobacteriaceae from food samples with this ready-to-use, selective medium.
Have Questions?
Catalog number PO5021A
Price (EUR)
-

Detect and isolate pathogenic Enterobacteriaceae from food samples with Thermo Scientific™ Oxoid™ Gassner Medium. It is a modification of the original Gassner (water blue metachrome yellow lactose) medium1. The medium contains metachrome yellow, which inhibits the Gram-positive accompanying flora. Lactose-digesting microorganisms such as Escherichia coli or Klebsiella species form blue colonies with simultaneous blue colouring of the surrounding agar. Lactose-negative microorganisms, such as Salmonella, Serratia or Proteus species, form yellow colonies with a yellow background.

Use Gassner Medium for selective detection and isolation of Enterobacteriaceae from food and other samples.

  • Selective: presence of metachrome yellow inhibits non-target Gram-positive flora.
  • Easy-to-read: Lactose positive organisms form blue colonies while lactose negative organisms form yellow colonies.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

For Laboratory Use
For Laboratory Use

1. Gassner, G. (1918), Zentralbl. Bakt. I. Orig., 80, 219-222.

Specifications
ColorOlive Green
Fill Weight17g ± 5%
Format90mm Monoplate
Quantity10/Pk.
Shelf Life26 Weeks
Manufacturing OriginGermany
Product TypeAgar Plate
Unit SizeEach
Contents & Storage
2°C to 10°C