iCAP™ PRO Series ICP-OES eLearning Courses
iCAP™ PRO Series ICP-OES eLearning Courses
Thermo Scientific™

iCAP™ PRO Series ICP-OES eLearning Courses

The iCAP™ PRO Series ICP-OES eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse TopicCourse LevelDuration
TICAPPROBOPBasic Operations4 to 8 hr.
TICAPROMTSUser Maintenance and TroubleshootingRoutine60 to 120 min.
Catalog number TICAPPROBOP
Price (EUR)
475,00
Each
Add to cart
Course Topic:
Basic Operations
Duration:
4 to 8 hr.
Price (EUR)
475,00
Each
Add to cart

The iCAP™ PRO Series ICP-OES eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The iCAP™ PRO Basic Operations eLearning course
Will provide the basics of using your instrument. It will cover the general theory behind how it works and the role of each component of the instrument. The course will also cover how to use the Qtegra software to set-up and run your methods and export the data. Included in the course are the following eLearning modules:

Thermo Scientific™ Qtegra™ ISDS Software:

  • Introduction to the Qtegra™ Software
  • Qtegra™ Dashboard
  • Creating a LabBook – Part 1
  • Creating a LabBook – Part 2
  • Reviewing and Exporting Data and Reports
  • Creating LabBook Templates

iCAP™ PRO ICP-OES:

  • Creating Calibration Standards Volumetrically
  • Startup and Shutdown Procedures
  • Concentric Glass Nebulizer
  • Cyclonic Spray Chamber
  • POP Window Maintenance
Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicBasic Operations
FormatSelf-paced eLearning
LanguagesEnglish
TechniqueInductively Couple Plasma – Optical Emission Spectroscopy
Time and DateOn-demand upon course end-user registration confirmation
Duration4 to 8 hr.
For Use With (Equipment)iCAP PRO Series ICP-OES
Product LineThermo Scientific
Product TypeAtomic Spectroscopy
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.