Search
Search
| Catalog Number | Capacity (English) | Door Style | Plug Type | Electrical Requirements |
|---|---|---|---|---|
| TSX5005SY | 51.1 cu. ft. | Double Solid | NEMA 5-15 | 115 V 60 Hz |
| TSX2305GA | 23 cu. ft. | Glass | NEMA 5-15 | 115V, 60Hz |
| TSX4505GA | 45.8 cu. ft. | Sliding Glass | NEMA 5-15 | 115V, 60Hz |
| TSX4505GD | 45.8 cu. ft. | Sliding Glass | NEMA 6-15 | 208-230V, 60Hz |
| TSX4505GV | 45.8 cu. ft. | Sliding Glass | CEE 7/7 | 208-230V, 50Hz |
| TSX5005GA | 51.1 cu. ft. | Double Glass | NEMA 5-15 | 115V, 60Hz |
| TSX5005GD | 51.1 cu. ft. | Double Glass | NEMA 6-15 | 208-230V, 60Hz |
| TSX5005GV | 51.1 cu. ft. | Double Glass | CEE 7/7 | 208-230V, 50Hz |
| TSX5005SA | 51.1 cu. ft. | Double Solid | NEMA 5-15 | 115V, 60Hz |
| TSX5005SD | 51.1 cu. ft. | Double Solid | NEMA 6-15 | 208-230V, 60Hz |
| TSX5005SV | 51.1 cu. ft. | Double Solid | CEE 7/7 | 208-230V, 50Hz |
| TSX1205GV | 11.5 cu. ft. | Glass | CEE 7/7 | 208-230V, 50Hz |
| TSX1205GY | 11.5 cu. ft. | Glass | NEMA 5-15 | 115 V 60 Hz |
| TSX1205GZ | 11.5 cu. ft. | Glass | NEMA 6-15 | 208 - 230 V 60 Hz |
| TSX1205SV | 11.5 cu. ft. | Solid | CEE 7/7 | 208-230V, 50Hz |
| TSX1205SY | 11.5 cu. ft. | Solid | NEMA 5-15 | 115 V 60 Hz |
| TSX1205SZ | 11.5 cu. ft. | Solid | NEMA 6-15 | 208 - 230 V 60 Hz |
| TSX3005GV | 29.2 cu. ft. | Glass | CEE 7/7 | 208-230V, 50Hz |
| TSX3005GY | 29.2 cu. ft. | Glass | NEMA 5-15 | 115 V 60 Hz |
| TSX3005GZ | 29.2 cu. ft. | Glass | NEMA 6-15 | 208 - 230 V 60 Hz |
| TSX3005SV | 29.2 cu. ft. | Solid | CEE 7/7 | 208-230V, 50Hz |
| TSX3005SY | 29.2 cu. ft. | Solid | NEMA 5-15 | 115 V 60 Hz |
| TSX3005SZ | 29.2 cu. ft. | Solid | NEMA 6-15 | 208 - 230 V 60 Hz |
| TSX5005GY | 51.1 cu. ft. | Double Glass | NEMA 5-15 | 115 V 60 Hz |
| TSX2305GD | 23 cu. ft. | Glass | NEMA 6-15 | 208-230V, 50/60Hz |
| TSX5005GZ | 51.1 cu. ft. | Double Glass | NEMA 6-15 | 208 - 230 V 60 Hz |
| TSX2305GV | 23 cu. ft. | Glass | CEE 7/7 | 208-230V, 50/60Hz |
| TSX2305SA | 23 cu. ft. | Solid | NEMA 5-15 | 115V, 60Hz |
| TSX5005SZ | 51.1 cu. ft. | Double Solid | NEMA 6-15 | 208 - 230 V 60 Hz |
| TSX2305SD | 23 cu. ft. | Solid | NEMA 6-15 | 208-230V, 50/60Hz |
| TSX2305GY | 23 cu. ft. | Glass | NEMA 5-15 | 115 V 60 Hz |
| TSX2305SV | 23 cu. ft. | Solid | CEE 7/7 | 208-230V, 50/60Hz |
| TSX2305GZ | 23 cu. ft. | Glass | NEMA 6-15 | 208 - 230 V 60 Hz |
| TSX2305SY | 23 cu. ft. | Solid | NEMA 5-15 | 115 V 60 Hz |
| TSX2305SZ | 23 cu. ft. | Solid | NEMA 6-15 | 208 - 230 V 60 Hz |
| TSX4505GY | 45.8 cu. ft. | Sliding Glass | NEMA 5-15 | 115 V 60 Hz |
| TSX4505GZ | 45.8 cu. ft. | Sliding Glass | NEMA 6-15 | 208 - 230 V 60 Hz |
Thermo Scientific™ TSX Series high-performance lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade storage requiring 3° to 7°C. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Featuring over 450W of reserve refrigeration capacity, the TSX series has the power to protect your product from temperature swings as a result of frequent and prolonged door access, large product loads, and other activities that introduce hear to the cabinet.
The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
The drive for sample integrity and security
Enabling compliance
Designed for an ergonomic and comfortable experience
Verified sustainability
Ready for connectivity and monitoring
Make it your own