Search
Grow a wide variety of microorganisms with Thermo Scientific™ Oxoid™ Tryptone Soya Broth ready-prepared medium. The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4and food testing methods for the growth of a wide variety of organisms.
Remove the need for autoclaving and dispensing with convenient, pre-filled and ready prepared tubes of Tryptone Soya Broth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
General References: