Fluorometric and Luminometric Filter Sets for Varioskan™ ALF Multimode Microplate Reader
Fluorometric and Luminometric Filter Sets for Varioskan™ ALF Multimode Microplate Reader
Fluorometric and Luminometric Filter Sets for Varioskan™ ALF Multimode Microplate Reader
Fluorometric and Luminometric Filter Sets for Varioskan™ ALF Multimode Microplate Reader
Thermo Scientific™

Fluorometric and Luminometric Filter Sets for Varioskan™ ALF Multimode Microplate Reader

Varioskan ALF Multimode Microplate Reader Filter Sets are high quality, validated, and available for various fluorometric and luminometric applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberExcitation/Emission
VAPAI012334/425 nm
VAPAI011452/545 nm
VAPAI007497/542 nm
VAPAI001535/595 nm
VAPAI005555/605 nm
VAPAI002625/677 nm
VAPAI003649/711 nm
VAPAI008390/513 nm
VAPAI009390/450 nm
VAPAI015334/450 nm
VAPAI006460 nm, 618 nm (Donor/Acceptor)
VAPAI010438/497 nm
VAPAI013527/615 nm
VAPAI014485/575 nm
VASET001438 nm, 497/542 nm (Donor/Acceptor)
VASET002485 nm, 525/595 nm (Donor/Acceptor)
VASET003535 nm, 595/677 nm (Donor/Acceptor)
VASET004390 nm, 450/525 nm (Donor/Acceptor)
Catalog number VAPAI012
Price (EUR)
902,00
Each
Excitation/Emission:
334/425 nm
Price (EUR)
902,00
Each
With the Varioskan ALF Multimode Microplate Reader, fluorescence and certain luminescence measurements are performed with filter-based optics for optimal specificity and sensitivity. The system comes with three pre-installed excitation and emission filter sets suitable for many common fluorometric applications like Quant-iT DNA quantitation assays and PrestoBlue and AlamarBlue cell viability assays. Additionally, five open filter pair positions are available for customized fluorometric and luminometric applications such as measurement of various Alexa Fluor™ and other common dyes or Promega NanoBRET™ assays. The filters are available as predefined, carefully tested, and validated filter sets, to ensure the optimal performance of selected assays.

Varioskan ALF instrument includes the following factory installed fluorometric filter pairs:

  • Coumarin family filter pair
    • Excitation 345 nm, emission 450 nm
  • Fluorescein family filter pair
    • Excitation 485, emission 525 nm
  • Resorufin family filter pair
    • Excitation 555 nm, emission 615 nm

In addition to factory installed filters, a range of validated filter sets are available for various fluorometric and luminometric applications.

  • Filter sets consists of high transmission filters for high excitation and emission efficiency.
  • Filter sets are designed to efficiently block unwanted light as well as unwanted emitted light to ensure high signal and low background.
  • Varioskan ALF filter installation is effortless
    • The instrument filter wheel is easy to access, and the correct parameters are quickly set in simple workflow in Thermo Scientific SkanIt software.
    • SkanIt software-controlled filter installation process prevents accidental mistakes ensuring error-free
For Laboratory Use
Specifications
Excitation/Emission334/425 nm
For Use With3-p-hydroxyphenyl-propionic acid (HPPA) assay and equivalent dyes
Plate Type6 to 384-well microplates
QuantityEach
For Use With (Equipment)Varioskan ALF Multimode Microplate Reader
Product TypeFilter Set
Unit SizeEach