Thermo Scientific Chemicals

Magnesium turnings, 1cm (0.4in) & down, Puratronic™, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 010630.14
25 g, Each
Thermo Scientific Chemicals

Magnesium turnings, 1cm (0.4in) & down, Puratronic™, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 010630.14
25 g, Each
Quantity

Chemical Identifiers

CAS
7439-95-4
IUPAC Name
magnesium
Molecular Formula
Mg
InChI Key
JLVVSXFLKOJNIY-UHFFFAOYSA-N
SMILES
[Mg]
Total Metal Impurities
0.02% max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable solids
    Category 1
    Self-heating substances/mixtures
    Category 2
    Substances/mixtures which, in contact with water, emit flammable gases
    Category 2
    Label Elements
    Signal Word

    Danger

    Physical Hazards

    H228 - Flammable solid

    H252 - Self-heating in large quantities; may catch fire

    H261 - In contact with water releases flammable gases

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P223 - Do not allow contact with water

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P335 + P334 - IF ON SKIN: Brush off loose particles from skin. Immerse in cool water

    P370 + P378 - In case of fire: Use fire-fighting equipment on basis class D for extinction

    P402 + P404 - Store in a dry place. Store in a closed container