Thermo Scientific Chemicals

Silver perchlorate monohydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 011624.04
2 g, Each
Thermo Scientific Chemicals

Silver perchlorate monohydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 011624.04
2 g, Each
Quantity
Catalog number: 011624.04
also known as 011624-04
Price (EUR)
Price: 30,00
Online price: 27,00
Your price:
Quantity
-

Chemical Identifiers

CAS
14242-05-8
IUPAC Name
silver(1+) perchlorate
Molecular Formula
AgClO4
InChI Key
YDHABVNRCBNRNZ-UHFFFAOYSA-M
SMILES
[Ag+].[O-][Cl](=O)(=O)=O
Total Metal Impurities
0.1% max.

Description

Silver perchlorate can be used in the manufacturing of explosives. It is generally used as a source of silver ions during silver nanoparticles synthesis. It is used as a catalyst in organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Oxidizing solids
    Category 2
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H272 - May intensify fire; oxidizer

    H314 - Causes severe skin burns and eye damage

    Precautionary Statements

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking