Magnesium perchlorate hexahydrate, 99% (metals basis)
Magnesium perchlorate hexahydrate, 99% (metals basis)
Magnesium perchlorate hexahydrate, 99% (metals basis)
Thermo Scientific Chemicals

Magnesium perchlorate hexahydrate, 99% (metals basis)

CAS: 13446-19-0 | Cl2H12MgO14 | 331.287 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 011635.A1
also known as 011635-A1
Price (EUR)
1.300,00
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
1.300,00
Each
Chemical Identifiers
CAS13446-19-0
IUPAC Namemagnesium(2+) hexahydrate diperchlorate
Molecular FormulaCl2H12MgO14
InChI KeyMGBDNCBRZIMSDZ-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Mg++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Commercially available magnesium perchlorate is reported as an extremely efficient catalyst for the synthesis of α-aminophosphonates. Magnesium perchlorate has been found to be an efficient catalyst for the synthesis of imines and phenylhydrazones by the reaction of carbonyl compounds with amines and phenylhydrazine in high yields at room temperatures and in short times.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only