Thermo Scientific Chemicals

Ammonium bromide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 012981.22
100 g, Each
Thermo Scientific Chemicals

Ammonium bromide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 012981.22
100 g, Each
Quantity
Catalog number: 012981.22
also known as 012981-22
Price (EUR)
Price: 371,00
Online price: 330,19
Your price:
Quantity
-

Chemical Identifiers

CAS
12124-97-9
IUPAC Name
ammonium bromide
Molecular Formula
BrH4N
InChI Key
SWLVFNYSXGMGBS-UHFFFAOYSA-N
SMILES
[NH4+].[Br-]
Total Metal Impurities
0.001% max.

Description

Ammonium bromide is used in pharmaceuticals for its bromide ion. It is used for a variety of analytical chemistry applications. It is also used for photography in films, plates and papers; in fireproofing of wood; in lithography and process engraving and as corrosion inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Serious Eye Damage/Eye Irritation
    Category 2
    Reproductive Toxicity
    Category 1B
    Specific target organ toxicity - (single exposure)
    Category 3
    Specific target organ toxicity - (repeated exposure)
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H319 - Causes serious eye irritation

    H336 - May cause drowsiness or dizziness

    H360FD - May damage fertility. May damage the unborn child

    H362 - May cause harm to breast-fed children

    H372 - Causes damage to organs through prolonged or repeated exposure

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    Additional EU labelling

    Restricted to professional users