Sodium hexafluorophosphate, 99+%
Sodium hexafluorophosphate, 99+%
Sodium hexafluorophosphate, 99+%
Sodium hexafluorophosphate, 99+%
Sodium hexafluorophosphate, 99+%
Thermo Scientific Chemicals

Sodium hexafluorophosphate, 99+%

CAS: 21324-39-0 | F6NaP | 167.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 013064.14
also known as 013064-14
Price (EUR)
93,50
Online Exclusive
110,00
Save 16,50 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
93,50
Online Exclusive
110,00
Save 16,50 (15%)
Each
Add to cart
Chemical Identifiers
CAS21324-39-0
IUPAC Namesodium hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6NaP
InChI KeyKMADQUOCJBLXRP-UHFFFAOYSA-N
SMILES[Na+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.1% min
Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only