Tin(II) sulfide, 99.5%
Tin(II) sulfide, 99.5%
Tin(II) sulfide, 99.5%
Thermo Scientific Chemicals

Tin(II) sulfide, 99.5%

CAS: 1314-95-0 | SSn | 150.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 014051.06
also known as 014051-06
Price (EUR)
150,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
150,00
Each
Chemical Identifiers
CAS1314-95-0
IUPAC Nameλ²-tin(2+) sulfanediide
Molecular FormulaSSn
InChI KeyDZXKSFDSPBRJPS-UHFFFAOYSA-N
SMILES[S--].[Sn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.5% min
Polymerization catalyst.Tin(II) sulfide is used as a polymerization catalyst. It is also used as a friction material. Further, it is used in optoelectronic device as well as a component in heterojunction photovoltaic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only