Thulium(III) nitrate hydrate, REacton™, 99.9% (REO)
Thulium(III) nitrate hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Thulium(III) nitrate hydrate, REacton™, 99.9% (REO)

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 014579.09
also known as 014579-09
Price (EUR)
293,25
Online Exclusive
345,00
Save 51,75 (15%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (EUR)
293,25
Online Exclusive
345,00
Save 51,75 (15%)
Each
Add to cart
Chemical Identifiers
CAS100641-15-4
IUPAC Namethulium(3+) trinitrate
Molecular FormulaN3O9Tm
InChI KeyLLZBVBSJCNUKLL-UHFFFAOYSA-N
SMILES[Tm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystalline aggregates
Total Rare Earth Oxide Impurities≤0.1% (≤1000ppm)
Total rare earth oxides≥99.9%
Thulium(III) nitrate hydrate is used as a laboratory reagent. It finds application in optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only