Praseodymium(III) acetate hydrate, 99.9% (REO)
Praseodymium(III) acetate hydrate, 99.9% (REO)
Praseodymium(III) acetate hydrate, 99.9% (REO)
Thermo Scientific Chemicals
Praseodymium(III) acetate hydrate, 99.9% (REO)
CAS: 334869-74-8 | C6H9O6Pr | 318.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 014611.14
also known as 014611-14
Price (EUR)
81,40
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
81,40
Each
Chemical Identifiers
CAS334869-74-8
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Praseodymium Acetate is used to make color glasses and enamels; when mixed with certain other materials, Praseodymium produces an intense clean yellow color in glass. The other main application is used to manufacturing pigment and ink color.