Silver carbonate is used in production of silver powder. It is used in microelectronics. Silver carbonate is used as a reagent in organic synthesis. It is employed in Fetizon oxidation, silver carbonate on celite acts as an oxidizing agent to form lactones from diols. It is also used in used in Koenigs-Knorr glycosylation. It is also employed to convert alkyl bromides into alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.