Sodium oxide
Sodium oxide
Sodium oxide
Thermo Scientific Chemicals

Sodium oxide

CAS: 1313-59-3 | Na2O | 61.979 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 036712.14
also known as 036712-14
Price (EUR)
87,55
Online Exclusive
103,00
Save 15,45 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
87,55
Online Exclusive
103,00
Save 15,45 (15%)
Each
Add to cart
Chemical Identifiers
CAS1313-59-3
IUPAC Namedisodium oxidandiide
Molecular FormulaNa2O
InChI KeyKKCBUQHMOMHUOY-UHFFFAOYSA-N
SMILES[O--].[Na+].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)78% min
Sodium oxide is used in chemical manufacturing, ceramics and glasses. Ethylbenzene dehydrogenation and carbon dioxide shift-reaction occurs using a sodium oxide/alumina catalyst. It is employed as catalyzing gasification of CO2 on carbon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only