Glassy carbon rod, 7mm (0.28in) dia, type 1
Thermo Scientific Chemicals

Glassy carbon rod, 7mm (0.28in) dia, type 1

Used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used as electrical conductor ion-selective membrane.
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mm
100 mm
200 mm
Catalog number 038012.DM
also known as 038012-DM
Price (EUR)
503,00
Each
Quantity:
100 mm
Request bulk or custom format
Price (EUR)
503,00
Each
Specifications
Chemical Name or MaterialGlassy carbon rod
EINECS Number231-153-3
MDL NumberMFCD00133992
PackagingTube
Recommended StorageAmbient temperatures
View more
Glassy carbon rod is used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used as electrical conductor ion-selective membrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the difference between type 1 glassy carbon and type 2 glassy carbon?

Type 1 is a "low temperature" glassy carbon produced by heat treatment at ~ 1,000-2,000°C and consists of discreet fragments of distorted graphene layers. In general, type 1 is less compressible, more resistant to oxidation, and less permeable to gasses. Type 2 is a"high temperature" glassy carbon produced by heat treatment at ~2,500-3,000°C and consists of broken or imperfect fullerene-like nanospheroids encased in a disordered, multilayered graphene matrix. Type 2 usually contains fewer impurities and has higher electrical and thermal conductivity.