Barium niobium oxide, 99.9% (metals basis)
Barium niobium oxide, 99.9% (metals basis)
Barium niobium oxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Barium niobium oxide, 99.9% (metals basis)

CAS: 12009-14-2 | BaNb2O6 | 419.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 039130.18
also known as 039130-18
Price (EUR)
275,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
275,00
Each
Chemical Identifiers
CAS12009-14-2
IUPAC Namebarium(2+) diniobiumoylolate
Molecular FormulaBaNb2O6
InChI KeyFKKSQMDUUDBSGH-UHFFFAOYSA-N
SMILES[Ba++].[O-][Nb](=O)=O.[O-][Nb](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities≤0.1%
Barium niobium oxide is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only