Lanthanum(III) nitride, 99.9% (REO)
Lanthanum(III) nitride, 99.9% (REO)
Lanthanum(III) nitride, 99.9% (REO)
Lanthanum(III) nitride, 99.9% (REO)
Thermo Scientific Chemicals

Lanthanum(III) nitride, 99.9% (REO)

CAS: 25764-10-7 | LaN | 152.912 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 039342.14
Price (EUR)
716,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
716,00
Each
Chemical Identifiers
CAS25764-10-7
IUPAC Namelanthanioamine
Molecular FormulaLaN
InChI KeyQCLQZCOGUCNIOC-UHFFFAOYSA-N
SMILESN#[La]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Lanthanum(III) nitride is used as a precursor along with silicon alkoxide to synthesize La9.33Si6O26, which finds application as electrolytes for solid oxide fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only